3-Bromopiridin-5-ol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Bromo-3-pyridinol, 3-Bromo-5-pyridinol, 5-Bromo-pyridin-3-ol
B3969-1G
B3969-5G
TCIAB3969-1GEA
31
EUR
TCIAB3969-1G
TCIAB3969-5G
3-Bromopiridin-5-ol ≥98.0% (by GC, titration analysis)
3-Bromopiridin-5-ol
Formula:
C₅H₄BrNO MW: 174 g/mol |
Número MDL:
MFCD00661305 Núm. CAS: 74115-13-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...