tert-Butyl (2-(methylamino)ethyl)carbamate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-Boc-N'-methyl-1,2-diaminoethane, tert-Butyl N-(2-Methylaminoethyl)carbamate, N-(2-Methylaminoethyl)carbamic acid tert-Butyl ester, N-Boc-N'-methylethylenediamine, N-(tert-Butoxycarbonyl)-N'-methylethylenediamine, N-(tert-Butoxycarbonyl)-N'-methyl-1,2-diaminoethane
B4046-5G
B4046-1G
TCIAB4046-5GEA
216
EUR
TCIAB4046-5G
TCIAB4046-1G
tert-Butyl (2-(methylamino)ethyl)carbamate ≥98.0% (by titrimetric analysis)
tert-Butyl (2-(methylamino)ethyl)carbamate
Formula:
C₈H₁₈N₂O₂ |
Número MDL:
MFCD06808585 Núm. CAS: 122734-32-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...