5-(2-Bromoethyl)-2,3-dihydrobenzofuran ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
5-(2-Bromoethyl)coumaran
B4066-5G
B4066-25G
TCIAB4066-5GEA
70
EUR
TCIAB4066-5G
TCIAB4066-25G
5-(2-Bromoethyl)-2,3-dihydrobenzofuran ≥98.0% (por GC)
5-(2-Bromoethyl)-2,3-dihydrobenzofuran
Formula:
C₁₀H₁₁BrO MW: 227,10 g/mol |
Número MDL:
MFCD06797641 Núm. CAS: 127264-14-6 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...