N-Benzylhydroxylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

B4073-5G B4073-25G
TCIAB4073-5GEA 119 EUR
TCIAB4073-5G TCIAB4073-25G
N-Benzylhydroxylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
N-Benzylhydroxylamine hydrochloride
Formula: C₇H₁₀ClNO
MW: 159.6134 g/mol
Número MDL: MFCD00043377
Núm. CAS: 29601-98-7

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 104.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR