2-(Benzylthio)nicotinic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(Benzylthio)pyridine-3-carboxylic Acid

B4088-1G B4088-5G
TCIAB4088-1GEA 53 EUR
TCIAB4088-1G TCIAB4088-5G
2-(Benzylthio)nicotinic acid ≥98.0% (by GC, titration analysis)
2-(Benzylthio)nicotinic acid
Formula: C₁₃H₁₁NO₂S
MW: 245,30 g/mol
Número MDL: MFCD00661380
Núm. CAS: 112811-90-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR