1-(4-Bromofenil)imidazol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(4-Bromofenil)-1H-imidazol
B4101-1G
B4101-5G
TCIAB4101-1GEA
67
EUR
TCIAB4101-1G
TCIAB4101-5G
1-(4-Bromofenil)imidazol ≥98.0% (by GC, titration analysis)
1-(4-Bromofenil)imidazol
Formula:
C₉H₇BrN₂ MW: 223,07 g/mol |
Número MDL:
MFCD00060489 Núm. CAS: 10040-96-7 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...