2-Bromobutyryl chloride ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
B4105-5G
B4105-25G
TCIAB4105-5GEA
125
EUR
TCIAB4105-5G
TCIAB4105-25G
2-Bromobutyryl chloride ≥98.0% (by GC, titration analysis)
2-Bromobutyryl chloride
Formula:
C₄H₆BrClO Pto. Ebullición: 65°C |
Número MDL:
MFCD00055269 Núm. CAS: 22118-12-3 EINECS: 244-791-2 UN: 2920 ADR: 8,II |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...