Brimonidine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: UK 14,304, 5-Bromo-N-(2-imidazolin-2-yl)-6-quinoxalinamine, 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine

B4132-200MG B4132-1G
TCIAB4132-200MGEA 18 EUR
TCIAB4132-200MG TCIAB4132-1G
Brimonidine ≥98.0% (by HPLC, titration analysis)
Brimonidine
Formula: C₁₁H₁₀BrN₅
MW: 292,14 g/mol
Número MDL: MFCD00153878
Núm. CAS: 59803-98-4
UN: 2811
ADR: 6.1,III
Índice Merck: 14,01375

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 245.0 to 249.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR