(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97.0% (por GC)
Proveedor: TCI
Sinónimos:
(2S,5S)-5-Benzyl-2-(tert-butyl)-3-methylimidazolidin-4-one, (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone
B4138-200MG
B4138-1G
TCIAB4138-200MGEA
79
EUR
TCIAB4138-200MG
TCIAB4138-1G
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97.0% (por GC)
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone
Formula:
C₁₅H₂₂N₂O MW: 246,35 g/mol |
Número MDL:
MFCD03426982 Núm. CAS: 346440-54-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 98.0 to 102.0 deg-C |
Specific rotation [a]20/D | -58.0 to -63.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...