N-(tert-Butoxycarbonyl)glycine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
Boc-Gly-OtBu, N-Boc-glycine tert-Butyl Ester
B4156-1G
B4156-5G
TCIAB4156-1GEA
24
EUR
TCIAB4156-1G
TCIAB4156-5G
N-(tert-Butoxycarbonyl)glycine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)glycine-tert-butyl ester
Formula:
C₁₁H₂₁NO₄ MW: 231,29 g/mol |
Número MDL:
MFCD00191866 Núm. CAS: 111652-20-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...