Bifonazole ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(alpha,4-Diphenylbenzyl)imidazole, 1-([1,1'-Biphenyl]-4-yl(phenyl)methyl)-1H-imidazole
B4173-5G
B4173-1G
TCIAB4173-5GEA
73
EUR
TCIAB4173-5G
TCIAB4173-1G
Bifonazole ≥98.0% (by GC, titration analysis)
Bifonazole
Formula:
C₂₂H₁₈N₂ MW: 310,40 g/mol |
Número MDL:
MFCD00865567 Núm. CAS: 60628-96-8 EINECS: 262-336-6 Índice Merck: 14,01213 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...