(Benzhydrylthio)acetic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(Benzhydrylthio)acetic acid, (Diphenylmethylthio)acetic Acid

B4175-5G B4175-25G
TCIAB4175-5GEA 80 EUR
TCIAB4175-5G TCIAB4175-25G
(Benzhydrylthio)acetic acid ≥98.0% (by GC, titration analysis)
(Benzhydrylthio)acetic acid
Formula: C₁₅H₁₄O₂S
MW: 258.34 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD05262040
Núm. CAS: 63547-22-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR