3-Bromo-5-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B4191-1G
B4191-5G
TCIAB4191-1GEA
32
EUR
TCIAB4191-1G
TCIAB4191-5G
3-Bromo-5-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-5-chlorobenzoic acid
Formula:
C₇H₄BrClO₂ MW: 235,46 g/mol |
Número MDL:
MFCD06797227 Núm. CAS: 42860-02-6 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...