3-Benciloxianilina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B4197-5G
B4197-25G
TCIAB4197-5GEA
43
EUR
TCIAB4197-5G
TCIAB4197-25G
3-Benciloxianilina ≥98.0% (by GC, titration analysis)
3-Benciloxianilina
Formula:
C₁₃H₁₃NO MW: 199,25 g/mol |
Número MDL:
MFCD00007784 Núm. CAS: 1484-26-0 EINECS: 216-056-6 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 63.0 to 68.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...