(R)-1-Boc-3-Hydroxypiperidine ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
(R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate
B4199-1G
B4199-5G
TCIAB4199-1GEA
45
EUR
TCIAB4199-1G
TCIAB4199-5G
(R)-1-Boc-3-Hydroxypiperidine ≥98.0% (por GC)
(R)-1-Boc-3-Hydroxypiperidine
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol |
Número MDL:
MFCD04115306 Núm. CAS: 143900-43-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Specific rotation [a]20/D | -20.0 to -23.0 deg (C=5,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...