(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B4202-200MG
B4202-1G
TCIAB4202-200MGEA
80
EUR
TCIAB4202-200MG
TCIAB4202-1G
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥98.0% (by GC, titration analysis)
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid
Formula:
C₁₀H₁₅F₂NO₄ |
Número MDL:
MFCD03094917 Núm. CAS: 203866-15-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -42.0 to -45.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...