2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥99.0% (by HPLC, total nitrogen), purified by sublimation
Proveedor: TCI
B4221-1G
B4221-5G
TCIAB4221-1GEA
81
EUR
TCIAB4221-1G
TCIAB4221-5G
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥99.0% (by HPLC, total nitrogen), purified by sublimation
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene
Formula:
C₂₆H₂₆N₂O₂S |
Número MDL:
MFCD00005774 Núm. CAS: 7128-64-5 EINECS: 230-426-4 Índice Merck: 14,01014 |
Resultados test especificaciones
Appearance | White to Light yellow to Green powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (with Total Nitrogen) | min. 99.0 % |
Melting point | 200.0 to 204.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...