tert-Butyl (2-aminophenyl)carbamate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-(tert-Butoxycarbonyl)-1,2-phenylenediamine, tert-Butyl (2-Aminophenyl)carbamate, 2-(tert-Butoxycarbonylamino)aniline, (2-Aminophenyl)carbamic acid tert-Butyl ester, 2-(Boc-amino)aniline, N-Boc-1,2-phenylenediamine
B4238-1G
B4238-5G
TCIAB4238-1GEA
46
EUR
TCIAB4238-1G
TCIAB4238-5G
tert-Butyl (2-aminophenyl)carbamate ≥98.0% (by HPLC, titration analysis)
tert-Butyl (2-aminophenyl)carbamate
Formula:
C₁₁H₁₆N₂O₂ MW: 208.2600 g/mol |
Número MDL:
MFCD02169707 Núm. CAS: 146651-75-4 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...