Bis(4-chloro-2-nitrophenyl)disulphide ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
B4247-5G
B4247-25G
TCIAB4247-5GEA
49
EUR
TCIAB4247-5G
TCIAB4247-25G
Bis(4-chloro-2-nitrophenyl)disulphide ≥98.0% (by HPLC, total nitrogen)
Bis(4-chloro-2-nitrophenyl)disulphide
Formula:
C₁₂H₆Cl₂N₂O₄S₂ |
Número MDL:
MFCD00047692 Núm. CAS: 2050-66-0 EINECS: 218-094-9 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 214.0 to 218.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...