1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene ≥97.0% (by HPLC)

Proveedor: TCI

B4252-200MG B4252-1G
TCIAB4252-200MGEA 44 EUR
TCIAB4252-200MG TCIAB4252-1G
1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene ≥97.0% (by HPLC)
1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene
Formula: C₃₀H₃₀N₄O₂
Número MDL: MFCD03093214
Núm. CAS: 138372-67-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 11.20 to 12.10 %
Melting point 239.0 to 243.0 deg-C

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR