N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester ≥98.0% (by HPLC)

Proveedor: TCI
Atención

Sinónimos: Methyl (tert-butoxycarbonyl)-L-phenylalaninate, N-Boc-L-phenylalanine Methyl Ester, Boc-Phe-OMe

B4259-5G B4259-25G
TCIAB4259-5GEA 24 EUR
TCIAB4259-5G TCIAB4259-25G
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester ≥98.0% (by HPLC)
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester
Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
Número MDL: MFCD00076977
Núm. CAS: 51987-73-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 37.0 to 41.0 deg-C
Specific rotation [a]20/D -3.0 to -5.0 deg (C=2, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR