4'-(Benzyloxy)benzylidene-4-fluoroaniline ≥98.0% (por GC)

Proveedor: TCI

B4301-5G B4301-25G
TCIAB4301-5GEA 45 EUR
TCIAB4301-5G TCIAB4301-25G
4'-(Benzyloxy)benzylidene-4-fluoroaniline ≥98.0% (por GC)
4'-(Benzyloxy)benzylidene-4-fluoroaniline
Formula: C₂₀H₁₆FNO
Número MDL: MFCD00017951
Núm. CAS: 70627-52-0

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR