Boc-L-norvaline ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Boc-Nva-OH, N-(tert-Butoxycarbonyl)-L-norvaline, (2S)-2-Aminopentanoic acid, N-BOC protected, N-Boc-L-norvaline

B4320-1G B4320-5G
TCIAB4320-1GEA 20 EUR
TCIAB4320-1G TCIAB4320-5G
Boc-L-norvaline ≥98.0% (by titrimetric analysis)
Boc-L-norvaline
Formula: C₁₀H₁₉NO₄
MW: 217,265 g/mol
Número MDL: MFCD00037268
Núm. CAS: 53308-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7ed7576e4b82f5165dec567f31116b2b0ce341d&vid=5f14b171e219bee9a36ebdd0d870051b8584de38&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 64.0 to 69.0 deg-C
Specific rotation [a]20/D -14.0 to -17.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR