(5-Bromopyridin-2-yl)hydrazine ≥96.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: (5-Bromo-2-pyridyl)hydrazine, 5-Bromo-2-hydrazinopyridine

B4327-1G B4327-5G
TCIAB4327-1GEA 30 EUR
TCIAB4327-1G TCIAB4327-5G
(5-Bromopyridin-2-yl)hydrazine ≥96.0% (by GC, titration analysis)
(5-Bromopyridin-2-yl)hydrazine
Formula: C₅H₆BrN₃
MW: 188,0252 g/mol
Número MDL: MFCD00513424
Núm. CAS: 77992-44-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Potassium iodate Method) min. 96.0 %
Melting point 132.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR