6-Bromo-4-quinazolone ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
6-Bromo-4(3H)-quinazolinone
B4328-5G
B4328-25G
TCIAB4328-5GEA
54
EUR
TCIAB4328-5G
TCIAB4328-25G
6-Bromo-4-quinazolone ≥98.0% (by GC, titration analysis)
6-Bromo-4-quinazolone
Formula:
C₈H₅BrN₂O MW: 225.04 g/mol |
Número MDL:
MFCD00115131 Núm. CAS: 32084-59-6 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...