N-(tert-Butoxycarbonyl)iminodiacetic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Boc-Ida-OH, Boc-DL-Ida-OH, N-Boc-iminodiacetic Acid

B4368-5G B4368-25G
TCIAB4368-5GEA 74 EUR
TCIAB4368-5G TCIAB4368-25G
N-(tert-Butoxycarbonyl)iminodiacetic acid ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)iminodiacetic acid
Formula: C₉H₁₅NO₆
MW: 233,22 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00800351
Núm. CAS: 56074-20-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR