1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
1-Benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride
B4393-5G
B4393-25G
TCIAB4393-5GEA
32
EUR
TCIAB4393-5G
TCIAB4393-25G
1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride ≥98.0% (by HPLC, titration analysis)
1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
Formula:
C₁₉H₂₁ClN₂ MW: 312,85 g/mol |
Número MDL:
MFCD00035361 Núm. CAS: 71258-18-9 EINECS: 275-305-7 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...