6-Bromopiridin-2-ol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Bromopiridin-6-ol

B4408-1G B4408-5G
TCIAB4408-1GEA 30 EUR
TCIAB4408-1G TCIAB4408-5G
6-Bromopiridin-2-ol ≥98.0% (by GC, titration analysis)
6-Bromopiridin-2-ol
Formula: C₅H₄BrNO
MW: 174 g/mol
Número MDL: MFCD00234043
Núm. CAS: 27992-32-1

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Purple powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR