5-Bromo-2-benzoxazolinone ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-Bromo-2-hydroxybenzoxazole, 5-Bromo-2-benzoxazolone, 5-Bromobenzo[d]oxazol-2(3H)-one

B4438-1G B4438-5G
TCIAB4438-1GEA 40 EUR
TCIAB4438-1G TCIAB4438-5G
5-Bromo-2-benzoxazolinone ≥98.0% (by GC, titration analysis)
5-Bromo-2-benzoxazolinone
Formula: C₇H₄BrNO₂
MW: 214,02 g/mol
Número MDL: MFCD01664246
Núm. CAS: 14733-73-4

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR