trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one ≥98.0% (by HPLC)
Proveedor: TCI
B4468-200MG
B4468-1G
TCIAB4468-200MGEA
67
EUR
TCIAB4468-200MG
TCIAB4468-1G
trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one ≥98.0% (by HPLC)
trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one
Formula:
C₁₉H₁₂F₆O |
Número MDL:
MFCD05662426 Núm. CAS: 103836-71-1 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...