N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
B4470-5G
B4470-25G
TCIAB4470-5GEA
75
EUR
TCIAB4470-5G
TCIAB4470-25G
N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine
Formula:
C₄₀H₃₆N₂O₂ MW: 576,74 g/mol |
Número MDL:
MFCD28384132 Núm. CAS: 169685-34-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...