1,3-Bis[4-(trifluoromethyl)phenyl]urea ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: 1,3-Bis(4-(trifluoromethyl)phenyl)urea

B4518-200MG B4518-1G
TCIAB4518-200MGEA 95 EUR
TCIAB4518-200MG TCIAB4518-1G
1,3-Bis[4-(trifluoromethyl)phenyl]urea ≥98.0% (by HPLC, total nitrogen)
1,3-Bis[4-(trifluoromethyl)phenyl]urea
Formula: C₁₅H₁₀F₆N₂O
MW: 348,25 g/mol
Número MDL: MFCD01908917
Núm. CAS: 1960-88-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR