N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
Methyl N,N'-Bis(tert-butoxycarbonyl)carbamimidothioate, N,N'-Di-Boc-carbamimidothioic Acid Methyl Ester, 1,3-Di-Boc-2-methylisothiourea, N,N'-Bis(tert-butoxycarbonyl)carbamimidothioic Acid Methyl Ester, N,N'-Di-Boc-S-methylisothiourea, Methyl N,N'-Di-Boc-carbamimidothioate, 1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea
B4535-5G
B4535-25G
TCIAB4535-5GEA
85
EUR
TCIAB4535-5G
TCIAB4535-25G
N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea ≥98.0% (by HPLC, total nitrogen)
N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea
Formula:
C₁₂H₂₂N₂O₄S MW: 290,38 g/mol |
Número MDL:
MFCD00239356 Núm. CAS: 107819-90-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...