2-Boc-aminothiazole-5-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 2-Boc-Aminothiazole-5-carboxylic acid, 2-(tert-Butoxycarbonylamino)thiazole-5-carboxylic acid, 2-(Boc-amino)thiazole-5-carboxylic acid

B4552-200MG B4552-1G
TCIAB4552-200MGEA 86 EUR
TCIAB4552-200MG TCIAB4552-1G
2-Boc-aminothiazole-5-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2-Boc-aminothiazole-5-carboxylic acid
Formula: C₉H₁₂N₂O₄S
MW: 244,2676 g/mol
Número MDL: MFCD06796614
Núm. CAS: 302964-02-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR