1,3-Bis(tert-butoxycarbonyl)thiourea ≥98.0% (by total nitrogen basis)
Proveedor: TCI
Sinónimos:
1,3-Di-Boc-thiourea
B4559-1G
B4559-5G
TCIAB4559-1GEA
140
EUR
TCIAB4559-1G
TCIAB4559-5G
1,3-Bis(tert-butoxycarbonyl)thiourea ≥98.0% (by total nitrogen basis)
1,3-Bis(tert-butoxycarbonyl)thiourea
Formula:
C₁₁H₂₀N₂O₄S MW: 276.35 g/mol |
Número MDL:
MFCD03093917 Núm. CAS: 145013-05-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...