O-tert-Butylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: O-(tert-Butyl)hydroxylamine hydrochloride

B4589-1G B4589-5G
TCIAB4589-1GEA 65 EUR
TCIAB4589-1G TCIAB4589-5G
O-tert-Butylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
O-tert-Butylhydroxylamine hydrochloride
Formula: C₄H₁₂ClNO
Número MDL: MFCD00043272
Núm. CAS: 39684-28-1
EINECS: 254-590-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR