1,3-Bis[4-(trifluoromethyl)phenyl]thiourea ≥97.0% (by HPLC, total nitrogen)

Proveedor: TCI

B4611-1G B4611-5G
TCIAB4611-1GEA 45 EUR
TCIAB4611-1G TCIAB4611-5G
1,3-Bis[4-(trifluoromethyl)phenyl]thiourea ≥97.0% (by HPLC, total nitrogen)
1,3-Bis[4-(trifluoromethyl)phenyl]thiourea
Formula: C₁₅H₁₀F₆N₂S
MW: 364,31 g/mol
Número MDL: MFCD18191886
Núm. CAS: 1744-07-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR