1,4-Benzenediboronic acid bis(pinacol)ester ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
1,4-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,4-Bis(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,4-Phenylenediboronic acid, pinacol ester
B4633-25G
B4633-5G
TCIAB4633-25GEA
306
EUR
TCIAB4633-25G
TCIAB4633-5G
1,4-Benzenediboronic acid bis(pinacol)ester ≥98.0% (por GC)
1,4-Benzenediboronic acid bis(pinacol)ester
Formula:
C₁₈H₂₈B₂O₄ MW: 330,04 g/mol |
Número MDL:
MFCD08276852 Núm. CAS: 99770-93-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 241.0 to 245.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...