Benzhydrylphosphonic acid ≥98.0% (by HPLC)
Proveedor: TCI
|
Peligro
|
B4634-200MG
B4634-1G
TCIAB4634-200MGEA
47
EUR
TCIAB4634-200MG
TCIAB4634-1G
Benzhydrylphosphonic acid ≥98.0% (by HPLC)
Benzhydrylphosphonic acid
Formula:
C₁₃H₁₃O₃P |
Número MDL:
MFCD00013952 Núm. CAS: 92025-81-5 UN: 3261 ADR: 8,II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...