4-Benzyloxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 4-Benzyloxy-2-fluorobenzeneboronic acid, (4-(Benzyloxy)-2-fluorophenyl)boronic acid

B4638-200MG B4638-1G
TCIAB4638-200MGEA 25 EUR
TCIAB4638-200MG TCIAB4638-1G
4-Benzyloxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Benzyloxy-2-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
MW: 246,04 g/mol
Número MDL: MFCD02093064
Núm. CAS: 166744-78-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR