Bis(9,9-dimethyl-9H-fluoren-2-yl)amine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

B4664-1G B4664-5G
TCIAB4664-1GEA 90 EUR
TCIAB4664-1G TCIAB4664-5G
Bis(9,9-dimethyl-9H-fluoren-2-yl)amine ≥98.0% (by HPLC, total nitrogen)
Bis(9,9-dimethyl-9H-fluoren-2-yl)amine
Formula: C₃₀H₂₇N
MW: 401,55 g/mol
Número MDL: MFCD23135317
Núm. CAS: 500717-23-7

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR