4′-Hydroxy-2-phenylacetophenone ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Benzyl 4-hydroxyphenyl ketone, 1-(4-Hydroxyphenyl)-2-phenylethanone
B4722-1G
B4722-5G
TCIAB4722-1GEA
41
EUR
TCIAB4722-1G
TCIAB4722-5G
4′-Hydroxy-2-phenylacetophenone ≥98.0% (by GC, titration analysis)
4′-Hydroxy-2-phenylacetophenone
Formula:
C₁₄H₁₂O₂ MW: 212,24 g/mol |
Número MDL:
MFCD00002360 Núm. CAS: 2491-32-9 EINECS: 219-654-5 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...