4-(Benzyloxycarbonyl)benzeneboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
(4-((Benzyloxy)carbonyl)phenyl)boronic acid, 4-Benzyloxycarbonylphenylboronic acid, 4-Carbobenzoxybenzeneboronic acid, 4-(Benzyloxycarbonyl)benzeneboronic acid, 4-Carbobenzoxyphenylboronic acid, 4-(Benzyloxycarbonyl)phenylboronic acid
B4725-1G
B4725-5G
TCIAB4725-1GEA
99
EUR
TCIAB4725-1G
TCIAB4725-5G
4-(Benzyloxycarbonyl)benzeneboronic acid (contains varying amounts of Anhydride)
4-(Benzyloxycarbonyl)benzeneboronic acid
Formula:
C₁₄H₁₃BO₄ MW: 256,06 g/mol |
Número MDL:
MFCD02179442 Núm. CAS: 184000-11-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...