Ácido 4-Butoxifenilborónico (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
(4-Butoxyphenyl)boronic acid, 4-n-Butoxyphenylboronic acid
B4727-1G
B4727-5G
TCIAB4727-1GEA
27
EUR
TCIAB4727-1G
TCIAB4727-5G
Ácido 4-Butoxifenilborónico (contains varying amounts of Anhydride)
Ácido 4-Butoxifenilborónico
Formula:
C₁₀H₁₅BO₃ MW: 194,04 g/mol |
Número MDL:
MFCD03427054 Núm. CAS: 105365-51-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...