Ácido (2-bromo-4-metoxifenil)acético ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(2-Bromo-4-methoxyphenyl)acetic acid, 2-Bromo-4-methoxyphenylacetic acid
B4737-1G
B4737-5G
TCIAB4737-1GEA
49
EUR
TCIAB4737-1G
TCIAB4737-5G
Ácido (2-bromo-4-metoxifenil)acético ≥98.0% (by GC, titration analysis)
Ácido (2-bromo-4-metoxifenil)acético
Formula:
C₉H₉BrO₃ MW: 245.07 g/mol |
Número MDL:
MFCD02664700 Núm. CAS: 66916-99-2 UN: 3077 ADR: 9 III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...