3-Bromo-2-etoxipiridina ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B4738-1G
B4738-5G
TCIAB4738-1GEA
65
EUR
TCIAB4738-1G
TCIAB4738-5G
3-Bromo-2-etoxipiridina ≥97.0% (by GC, titration analysis)
3-Bromo-2-etoxipiridina
Formula:
C₇H₈BrNO MW: 202,05 g/mol |
Número MDL:
MFCD00234309 Núm. CAS: 57883-25-7 |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...