2-(3-Bromophenyl)triphenylene ≥98.0% (por GC)

Proveedor: TCI

B4746-200MG B4746-1G
TCIAB4746-200MGEA 22 EUR
TCIAB4746-200MG TCIAB4746-1G
2-(3-Bromophenyl)triphenylene ≥98.0% (por GC)
2-(3-Bromophenyl)triphenylene
Formula: C₂₄H₁₅Br
MW: 383,29 g/mol
Número MDL: MFCD28384139
Núm. CAS: 1313514-53-2

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR