1-(4-Bromophenyl)-1,3-butanedione ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
(4-Bromobenzoyl)acetone, 1-(4-Bromophenyl)butane-1,3-dione
B4754-1G
B4754-5G
TCIAB4754-1GEA
143
EUR
TCIAB4754-1G
TCIAB4754-5G
1-(4-Bromophenyl)-1,3-butanedione ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)-1,3-butanedione
Formula:
C₁₀H₉BrO₂ MW: 241,08 g/mol |
Número MDL:
MFCD04619579 Núm. CAS: 4023-81-8 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...