(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
N-(tert-Butoxycarbonyl)-D-proline methyll ester, Boc-D-Pro-OMe, N-Boc-D-proline methyll ester, N-Boc-D-proline methyl ester
B4787-1G
B4787-5G
TCIAB4787-1GEA
17
EUR
TCIAB4787-1G
TCIAB4787-5G
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate ≥98.0% (por GC)
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate
Formula:
C₁₁H₁₉NO₄ MW: 229.27 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00190828 Núm. CAS: 73323-65-6 |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid to cloudy liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +64.0 to +67.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...