N,N'-Bis(4-formylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)

Proveedor: TCI

B4793-200MG B4793-1G
TCIAB4793-200MGEA 39 EUR
TCIAB4793-200MG TCIAB4793-1G
N,N'-Bis(4-formylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)
N,N'-Bis(4-formylphenyl)-N,N'-diphenylbenzidine
Formula: C₃₈H₂₈N₂O₂
Número MDL: MFCD03093284
Núm. CAS: 137911-28-5

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR